
2-Acetamido-2-deoxy-4,6-O-isopropylidene-D-glucopyranose
Catalog # APOSOR5835T-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:184…186 °C (dec.)
:261.27 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:50605-09-9
:564469-85-8
Formula:C₁₁H₁₉NO₆
MDL Number:MFCD09260464