Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dibromo-1H-pyrrole-2-carboxaldehyde
4,5-Dibromo-1H-pyrrole-2-carboxaldehyde 98%
4,5-Dibromo-1H-pyrrole-2-carboxaldehyde 98%
APOSOR5763-1G
:
Apollo Scientific
:
4,5-Dibromo-1H-pyrrole-2-carboxaldehyde 98%
APOSOR5763-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
252.891 g/mol
:
154…158 °C
:
GHS07
:
Refrigerator
:
932-82-1
:
C₅H₃Br₂NO
:
MFCD07363813