Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4-methoxybenzene-1,2-diamine
3-Bromo-4-methoxybenzene-1,2-diamine 95%
3-Bromo-4-methoxybenzene-1,2-diamine 95%
3-Bromo-4-methoxybenzene-1,2-diamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-4-methoxybenzene-1,2-diamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1831097-76-7
:
Formula:
C₇H₉BrN₂O
MDL Number:
MFCD30685009
:
217.063 g/mol
Storage Temperature:
Ambient