Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methoxy-1,3-benzothiazole
5-Methoxy-1,3-benzothiazole 95%
5-Methoxy-1,3-benzothiazole 95%
Irritant
5-Methoxy-1,3-benzothiazole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Methoxy-1,3-benzothiazole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2942-14-5
:
Formula:
C₈H₇NOS
MDL Number:
MFCD06659660
:
165.212 g/mol
:
GHS07
Storage Temperature:
Refrigerator