Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dibromo-2,3-dichlorobenzene
1,4-Dibromo-2,3-dichlorobenzene 95%
1,4-Dibromo-2,3-dichlorobenzene 95%
Irritant
1,4-Dibromo-2,3-dichlorobenzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,4-Dibromo-2,3-dichlorobenzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
100191-20-6
:
Formula:
C₆H₂Br₂Cl₂
MDL Number:
MFCD09878188
:
304.794 g/mol
:
GHS07
Storage Temperature:
Ambient