Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Isopropoxybenzene-1,3-diboronic acid
4-Isopropoxybenzene-1,3-diboronic acid 98%
4-Isopropoxybenzene-1,3-diboronic acid 98%
Irritant
4-Isopropoxybenzene-1,3-diboronic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Isopropoxybenzene-1,3-diboronic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
850568-40-0
:
Formula:
C₉H₁₄B₂O₅
MDL Number:
MFCD04115638
:
223.826 g/mol
Melting Pt:
104…108 °C
:
GHS07
Storage Temperature:
Refrigerator