Order Entry
Export
ContactUsLinkComponent
2,4-Dichloro-5-cyclopropylpyrimidine 95%
2,4-Dichloro-5-cyclopropylpyrimidine 95%
Catalog # APOSOR55311-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloro-5-cyclopropylpyrimidine 95%
Catalog # APOSOR55311-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.042 g/mol
Storage Temperature:Ambient
CAS Number:1190379-86-2
Formula:C₇H₆Cl₂N₂
MDL Number:MFCD18533532