Order Entry
Export
ContactUsLinkComponent
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%
Irritant
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%
Catalog # APOSOR55309-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%
Catalog # APOSOR55309-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.262 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:869494-16-6
Formula:C₁₀H₁₈N₂O₂
MDL Number:MFCD17016736