

2,3-Dimethoxy-2,3-dimethylbutane
Catalog # APOSOR55306-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
ADR:3,III
:146.227 g/mol
UN:1993
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:6091-59-4
Formula:C₈H₁₈O₂
MDL Number:MFCD28137218