1,4-Diaminonaphthalene
Catalog # APOSOR55288-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
Melting Pt:114…116 °C
:158.2 g/mol
Storage Temperature:Ambient
CAS Number:2243-61-0
Formula:C₁₀H₁₀N₂
MDL Number:MFCD01712813