
2-Azaspiro[3.4]octane hemioxalate
Catalog # APOSOR55128-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:312.405 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1523617-94-8
Formula:2C₇H₁₃N·C₂H₂O₄
MDL Number:MFCD27988100