Order Entry
Export
ContactUsLinkComponent
Diethyl cyclopropane-1,2-dicarboxylate 95%
Diethyl cyclopropane-1,2-dicarboxylate 95%
Catalog # APOSOR55008-1G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl cyclopropane-1,2-dicarboxylate 95%
Catalog # APOSOR55008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.205 g/mol
Storage Temperature:Ambient
CAS Number:20561-09-5
Formula:C₉H₁₄O₄
MDL Number:MFCD00001286