Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dimethyloctane

2,4-Dimethyloctane

Flammable
Irritant
Supplier:   Apollo Scientific
2,4-Dimethyloctane
Supplier:  Apollo Scientific
Certificates
2,4-Dimethyloctane

2 Options Available Below

Product Details & Documents
  • CAS Number:4032-94-4
  • Formula:C₁₀H₂₂
  • MDL Number:MFCD30534470
  • Molecular Weight:142.282 g/mol
  • Boiling Pt:152…153 °C
  • Storage Temperature:Ambient
  • UN:3295
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 139,00 €
    139,00 €
    1 * 250 mg
       139,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 371,00 €
    371,00 €
    1 * 1 g
       371,00 €