2,3-Dihydroxy-4-methoxybenzoic acid
Catalog # APOSOR54556-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.146 g/mol
Storage Temperature:Ambient
CAS Number:3934-81-4
Formula:C₈H₈O₅
MDL Number:MFCD00016499