0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,5-Dichloropentan-3-one
1,5-Dichloropentan-3-one
1,5-Dichloropentan-3-one
:
Apollo Scientific
1,5-Dichloropentan-3-one
:
Apollo Scientific
:
3592-25-4
:
:
C₅H₈Cl₂O
:
MFCD06659623
:
155.022 g/mol
:
GHS07
:
Refrigerator