5-Iodo-1,2,3,4-tetrahydro-1,8-naphthyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:945600-06-6
:
Formula:C₈H₉IN₂
MDL Number:MFCD22548134
:260.075 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: