Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
1256162-94-3
:
Formula:
C₇H₄ClN₃O
MDL Number:
MFCD22551530
:
181.579 g/mol
Storage Temperature:
Ambient