Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Dicyano-4-phenylbutadiene
1,1-Dicyano-4-phenylbutadiene 98%
1,1-Dicyano-4-phenylbutadiene 98%
1,1-Dicyano-4-phenylbutadiene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
1,1-Dicyano-4-phenylbutadiene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
5439-39-4
:
Formula:
C₁₂H₈N₂
MDL Number:
MFCD01578804
:
180.205 g/mol
:
GHS07
Storage Temperature:
Ambient