Order Entry
Export
ContactUsLinkComponent
3,3-Dimethyloxan-4-amine hydrochloride
3,3-Dimethyloxan-4-amine hydrochloride
Catalog # APOSOR52315-1G
Supplier:  Apollo Scientific
CAS Number:  
3,3-Dimethyloxan-4-amine hydrochloride
Catalog # APOSOR52315-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :165.661 g/mol
Storage Temperature:Refrigerator
CAS Number:1400580-54-2
Formula:C₇H₁₅NO·ClH
MDL Number:MFCD28155170