

2-Methyl-4-[(methylamino)methyl]-1,3-thiazole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:144163-81-5
:
Formula:C₆H₁₀N₂S
MDL Number:MFCD06738964
:142.222 g/mol
UN:2922
ADR:8+6.1,III
:GHS05
:GHS07
Storage Temperature:Ambient