Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Iodo-1H-pyrazolo[3,4-c]pyridine
3-Iodo-1H-pyrazolo[3,4-c]pyridine 98%
3-Iodo-1H-pyrazolo[3,4-c]pyridine 98%
:
Apollo Scientific
3-Iodo-1H-pyrazolo[3,4-c]pyridine 98%
:
Apollo Scientific
:
1082040-63-8
:
:
C₆H₄IN₃
:
MFCD11845480
:
245.021 g/mol
:
GHS07
:
Refrigerator