Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dichlorophenylacetylene

3,4-Dichlorophenylacetylene

Irritant
Supplier:   Apollo Scientific
3,4-Dichlorophenylacetylene
Supplier:  Apollo Scientific
Certificates
3,4-Dichlorophenylacetylene

4 Options Available Below

Product Details & Documents
Synonyms:  1,2-Dichloro-4-ethynylbenzene
  • CAS Number:556112-20-0
  • Formula:C₈H₄Cl₂
  • MDL Number:MFCD07783046
  • Molecular Weight:171.023 g/mol
  • Melting Pt:40…44 °C
  • Flash Pt:96.1 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 53,60 €
    53,60 €
    1 * 1 g
       53,60 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 66,60 €
    66,60 €
    1 * 5 g
       66,60 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 278,00 €
    278,00 €
    1 * 25 g
       278,00 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 834,00 €
    834,00 €
    1 * 100 g
       834,00 €