Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,7-Dibromobenzo[d]thiazole
5,7-Dibromobenzo[d]thiazole ≥98%
5,7-Dibromobenzo[d]thiazole ≥98%
5,7-Dibromobenzo[d]thiazole ≥98%
APOSOR510263-100MG
:
Apollo Scientific
:
5,7-Dibromobenzo[d]thiazole ≥98%
APOSOR510263-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
292.978 g/mol
:
Ambient
:
875-69-4
:
C₇H₃Br₂NS
:
MFCD12962849