Order Entry
Export
ContactUsLinkComponent
 
1,7-Naphthyridine-3-carboxylic acid ≥98%
Catalog # APOSOR510245-100MG
Supplier:  Apollo Scientific
CAS Number:  
1,7-Naphthyridine-3-carboxylic acid ≥98%
Catalog # APOSOR510245-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :174.156 g/mol
Storage Temperature:Ambient
CAS Number:250674-49-8
Formula:C₉H₆N₂O₂
MDL Number:MFCD11044184