Order Entry
Export
ContactUsLinkComponent
2-Bromo-5-iodoimidazo[2,1-b]thiazole ≥98%
2-Bromo-5-iodoimidazo[2,1-b]thiazole ≥98%
Catalog # APOSOR510240-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-5-iodoimidazo[2,1-b]thiazole ≥98%
Catalog # APOSOR510240-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :328.956 g/mol
Storage Temperature:Ambient
CAS Number:1379307-48-8
Formula:C₅H₂BrIN₂S