Order Entry
Export
ContactUsLinkComponent
6-Bromo-[1,2,4]triazolo[4,3-a]pyrimidine ≥98%
6-Bromo-[1,2,4]triazolo[4,3-a]pyrimidine ≥98%
Catalog # APOSOR510235-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-[1,2,4]triazolo[4,3-a]pyrimidine ≥98%
Catalog # APOSOR510235-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :199.008 g/mol
Storage Temperature:Ambient
CAS Number:1019023-12-1
Formula:C₅H₃BrN₄
MDL Number:MFCD09994391