4,6-Dichloro-3-iodo-1H-pyrazolo[3,4-d]pyrimidine 95%
Catalog # APOSOR510192-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:314.899 g/mol
Storage Temperature:Refrigerator
CAS Number:1379308-33-4
Formula:C₅HCl₂IN₄
MDL Number:MFCD20258005