Order Entry
Export
ContactUsLinkComponent
6-Benzyl-2-oxa-6-azaspiro[3.3]heptane ≥98%
6-Benzyl-2-oxa-6-azaspiro[3.3]heptane ≥98%
Catalog # APOSOR510151-100MG
Supplier:  Apollo Scientific
CAS Number:  
6-Benzyl-2-oxa-6-azaspiro[3.3]heptane ≥98%
Catalog # APOSOR510151-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :189.254 g/mol
Storage Temperature:Ambient
CAS Number:46246-91-7
Formula:C₁₂H₁₅NO
MDL Number:MFCD12407094