Order Entry
Export
ContactUsLinkComponent
4-Chlorothieno[2,3-b]pyridine ≥98%
4-Chlorothieno[2,3-b]pyridine ≥98%
Catalog # APOSOR510139-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Chlorothieno[2,3-b]pyridine ≥98%
Catalog # APOSOR510139-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :169.631 g/mol
Storage Temperature:Ambient
CAS Number:62226-17-9
Formula:C₇H₄ClNS
MDL Number:MFCD11858513