2,3-Dihydrofuro[2,3-b]pyridin-3-amine ≥98%
Catalog # APOSOR510130-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:136.151 g/mol
Storage Temperature:Ambient
CAS Number:870061-94-2
Formula:C₇H₈N₂O
MDL Number:MFCD13190275