Order Entry
Export
ContactUsLinkComponent
Ethyl 6-bromo-4,8-dichloroquinoline-3-carboxylate > 95%
Ethyl 6-bromo-4,8-dichloroquinoline-3-carboxylate > 95%
Catalog # APOSOR510119-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6-bromo-4,8-dichloroquinoline-3-carboxylate > 95%
Catalog # APOSOR510119-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :349.007 g/mol
Storage Temperature:Freezer
CAS Number:1019396-10-1
Formula:C₁₂H₈BrCl₂NO₂
MDL Number:MFCD11522695