Order Entry
Export
ContactUsLinkComponent
 
3-Amino-4-bromothiophene-2-carboxylic acid ≥98%
Catalog # APOSOR510069-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-4-bromothiophene-2-carboxylic acid ≥98%
Catalog # APOSOR510069-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :222.06 g/mol
Storage Temperature:Refrigerator
CAS Number:215927-34-7
Formula:C₅H₄BrNO₂S