Order Entry
Export
ContactUsLinkComponent
2-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid ≥95%
2-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid ≥95%
Catalog # APOSOR510063-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid ≥95%
Catalog # APOSOR510063-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :196.59 g/mol
Storage Temperature:Refrigerator
CAS Number:522604-25-7
Formula:C₈H₅ClN₂O₂