Order Entry
Export
ContactUsLinkComponent
3-Chloro-2-iodo-5,6-dimethoxypyridine ≥98%
3-Chloro-2-iodo-5,6-dimethoxypyridine ≥98%
Catalog # APOSOR510053-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-2-iodo-5,6-dimethoxypyridine ≥98%
Catalog # APOSOR510053-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :299.493 g/mol
Storage Temperature:Refrigerator
CAS Number:910616-72-7
Formula:C₇H₇ClINO₂
MDL Number:MFCD22056183