Order Entry
Export
ContactUsLinkComponent
6-Bromo-7-chloroimidazo[1,2-a]pyridine ≥97%
6-Bromo-7-chloroimidazo[1,2-a]pyridine ≥97%
Catalog # APOSOR510043-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-7-chloroimidazo[1,2-a]pyridine ≥97%
Catalog # APOSOR510043-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :231.477 g/mol
Storage Temperature:Ambient
CAS Number:1303890-45-0
Formula:C₇H₄BrClN₂
MDL Number:MFCD12405308