Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde ≥97%
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde ≥97%
APOSOR510039-1G
:
Apollo Scientific
:
5-Bromobenzo[d][1,3]dioxole-4-carbaldehyde ≥97%
APOSOR510039-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
229.027 g/mol
:
Refrigerator
:
72744-54-8
:
C₈H₅BrO₃
:
MFCD02069012