Order Entry
Export
ContactUsLinkComponent
 
4-Bromopyrazolo[1,5-a]pyridine ≥98%
Catalog # APOSOR510024-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromopyrazolo[1,5-a]pyridine ≥98%
Catalog # APOSOR510024-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :197.032 g/mol
Storage Temperature:Ambient
CAS Number:1427404-87-2
Formula:C₇H₅BrN₂
MDL Number:MFCD23710563