Order Entry
Export
ContactUsLinkComponent
5-Chloroimidazo[1,2-a]pyridine-2-carbaldehyde ≥97%
5-Chloroimidazo[1,2-a]pyridine-2-carbaldehyde ≥97%
Catalog # APOSOR510018-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Chloroimidazo[1,2-a]pyridine-2-carbaldehyde ≥97%
Catalog # APOSOR510018-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :180.591 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
CAS Number:881841-35-6
Formula:C₈H₅ClN₂O