Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Bromo-1,7-dihydropurin-6-one
8-Bromo-1,7-dihydropurin-6-one
8-Bromo-1,7-dihydropurin-6-one
Irritant
8-Bromo-1,7-dihydropurin-6-one
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
8-Bromo-1,7-dihydropurin-6-one
Supplier:
Apollo Scientific
About this item
CAS Number:
56046-36-7
:
Formula:
C₅H₃BrN₄O
MDL Number:
MFCD01317991
:
215.008 g/mol
:
GHS07
Storage Temperature:
Ambient