Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-1,7-naphthyridin-8(7H)-one
3-Bromo-1,7-naphthyridin-8(7H)-one 97%
3-Bromo-1,7-naphthyridin-8(7H)-one 97%
3-Bromo-1,7-naphthyridin-8(7H)-one 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-1,7-naphthyridin-8(7H)-one 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
1375301-90-8
:
Formula:
C₈H₅BrN₂O
MDL Number:
MFCD22548037
:
225.042 g/mol
:
GHS07
Storage Temperature:
Ambient