Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromoimidazo[1,2-a]pyrimidine
7-Bromoimidazo[1,2-a]pyrimidine 97%
7-Bromoimidazo[1,2-a]pyrimidine 97%
7-Bromoimidazo[1,2-a]pyrimidine 97%
APOSOR500037-250MG
:
Apollo Scientific
:
7-Bromoimidazo[1,2-a]pyrimidine 97%
APOSOR500037-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
198.02 g/mol
:
GHS07
:
Freezer
:
1251033-57-4
:
C₆H₄BrN₃
:
MFCD16877232