Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2H-pyrazolo[4,3-c]pyridine
4-Chloro-2H-pyrazolo[4,3-c]pyridine 95+%
4-Chloro-2H-pyrazolo[4,3-c]pyridine 95+%
Irritant
4-Chloro-2H-pyrazolo[4,3-c]pyridine 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Chloro-2H-pyrazolo[4,3-c]pyridine 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
1821250-10-5
:
Formula:
C₆H₄ClN₃
MDL Number:
MFCD31568525
:
153.569 g/mol
:
GHS07
Storage Temperature:
Ambient