
Methyl 4,6-dibromopicolinate 95+%
Catalog # APOSOR500014-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:294.928 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1206248-47-6
Formula:C₇H₅Br₂NO₂
MDL Number:MFCD14582014