
4-Amino-2,5-dibromophenol 95+%
Catalog # APOSOR500009-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:266.918 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:872272-33-8
Formula:C₆H₅Br₂NO
MDL Number:MFCD28040627