Imidazo[1,5-a]pyrazin-8-amine
Catalog # APOSOR49048-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:134.139 g/mol
Storage Temperature:Ambient
CAS Number:26538-77-2
Formula:C₆H₆N₄
MDL Number:MFCD09835041