Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Chlorothieno[2,3-c]pyridine
7-Chlorothieno[2,3-c]pyridine 95%
7-Chlorothieno[2,3-c]pyridine 95%
Irritant
7-Chlorothieno[2,3-c]pyridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
7-Chlorothieno[2,3-c]pyridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
28948-58-5
:
Formula:
C₇H₄ClNS
MDL Number:
MFCD04973300
Melting Pt:
39…42 °C
:
169.631 g/mol
:
GHS07
Storage Temperature:
Ambient