0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Tetrahydropyran-4,4-dicarbonitrile
Tetrahydropyran-4,4-dicarbonitrile 95%
Tetrahydropyran-4,4-dicarbonitrile 95%
Tetrahydropyran-4,4-dicarbonitrile 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Tetrahydropyran-4,4-dicarbonitrile 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
111511-90-1
:
Formula:
C₇H₈N₂O
MDL Number:
MFCD19313175
:
136.151 g/mol
Storage Temperature:
Ambient