2-Bromo-4,5-dimethylaniline 95%
Catalog # APOSOR480643-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:200.076 g/mol
Storage Temperature:Ambient
CAS Number:22364-29-0
Formula:C₈H₁₀BrN
MDL Number:MFCD00031416