2-(p-tolyl)-1,3-benzothiazole 98+%
Catalog # APOSOR480550-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:225.309 g/mol
Storage Temperature:Ambient
CAS Number:16112-21-3
Formula:C₁₄H₁₁NS
MDL Number:MFCD00030228