Order Entry
Export
ContactUsLinkComponent
Ethyl 5-amino-1-(2-cyanoethyl)-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-cyanoethyl)-4-iodo-pyrazole-3-carboxylate
Catalog # APOSOR480031-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5-amino-1-(2-cyanoethyl)-4-iodo-pyrazole-3-carboxylate
Catalog # APOSOR480031-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :334.114 g/mol
Storage Temperature:Ambient
CAS Number:1427014-07-0
Formula:C₉H₁₁IN₄O₂
MDL Number:MFCD23135197